Learn More
MilliporeSigma™ Supelco™ Hydrocarbon Kit acc. to DIN EN 14039/ISO 16703
Catalog No. 111016458 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Hydrocarbon Kit acc. to DIN EN 14039/ISO 16703
Catalog No.
11-101-6458 Hydrocarbon Kit acc. to DIN EN 14039/ISO 16703
1 options
Catalog No. 11-101-6458 Supplier MilliporeSigma™ Supelco™ Supplier No. 566811KTF
Add to Cart
Add to Cart
Kit contains:
  • mineral oil standard for calibration 8 mg/ml in heptane
  • alkane standard solution
  • heptane
  • standard solution for the determination of the retention time
  • Florisil™ tubes
TRUSTED_SUSTAINABILITY
Specifications
Description Limited shelf life, expiry date on the label