Learn More
Ideal Vacuum Products LLC Alcatel Adixen A119 Hydrocarbon Vacuum Pump Oil, 4 Liter
SDP
Catalog No. NC1620784
Encompass
Alcatel Adixen A119 Hydrocarbon Vacuum Pump Oil, 4 Liter (~1 Gallon) 98102 is quality mechanical vacuum pump oil for the ultimate performance of your Alcatel Rotary Vane pumps. Alcatel Adixen A119 Hydrocarbon Oil is specially prepared by Alcatel Adixen to meet the Rotary Vane and RSV Series Boost Pumps exclusive specifications. We recommend that you change your vacuum pump oil at least every 3 months to prolong the life of your vacuum pump.
Catalog No. NC1620784 Supplier Ideal Vacuum Products LLC Supplier No. P102102
Alcatel Adixen A119 Hydrocarbon Vacuum Pump Oil, 4 Liter (~1 Gallon) 98102 is quality mechanical vacuum pump oil for the ultimate performance of your Alcatel Rotary Vane pumps. Alcatel Adixen A119 Hydrocarbon Oil is specially prepared by Alcatel Adixen to meet the Rotary Vane and RSV Series Boost Pumps exclusive specifications. We recommend that you change your vacuum pump oil at least every 3 months to prolong the life of your vacuum pump.