Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.0100 N (N/100), Ricca Chemical
SDP

Catalog No. 35901 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
35901 4 L Natural Poly Bottle
3590-4 120 mL Natural Poly Bottle
3590-32 1 L Natural Poly Bottle
R359000010F 10 L Cubitainer
35905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 35901 Supplier Ricca Chemical Company Supplier No. 35901
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance of Solution Colorless Liquid
Concentration 0.00995 to 0.01005 N
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Quantity 4 L
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0100 N
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.