Learn More
Electron Microscopy Sciences Hydrochloric Acid 0.01n 500 ML
SDP
Catalog No. 5030075
Encompass_Preferred
Used in Picrosirius Red Staining of Cardiac muscle following phosphomolybdic acid treatment. For connective tissue stains – Van Gieson stains and variants. See Dolber, Paul C. and Madison S. Spach. Picrosirus Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment, Stain Technology, Vol. 62 No 1 (1987) 23 – 26.
Catalog No. 50-300-75 Supplier Electron Microscopy Sciences Supplier No. 2635703
Used in Picrosirius Red Staining of Cardiac muscle following phosphomolybdic acid treatment. For connective tissue stains – Van Gieson stains and variants. See Dolber, Paul C. and Madison S. Spach. Picrosirus Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment, Stain Technology, Vol. 62 No 1 (1987) 23 – 26.