Learn More
Hach Company Hydrochloric Acid Standard Solution, 0.02 N, 1 L
Supplier:  Hach Company 2330353
Encompass
For determination of alkalinity by buret titration. APHA formulation. 1 liter bottle.
Catalog No. 50-199-5822