Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.0500 N (N/20), Ricca Tightrant™
SDP

Catalog No. R35970004F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R35970004F 4 L Cubitainer
R3597005250 250 mL Natural Poly Bottle
R3597000500 500 mL Natural Poly Bottle
3597-32 1 L Natural Poly Bottle
3597-1 4 L Natural Poly Bottle
3597-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R35970004F Supplier Ricca Chemical Company Supplier No. 35971CT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid Titrant

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Concentration 0.0500Normal
CAS Min % 98.0, 0.48
CAS Max % 100.0, 0.5
Packaging Cubitainer
Quantity 4 L
Physical Form Liquid
Grade Not Available
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.