Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.0500 N (N/20), Ricca Tightrant™
SDP

Catalog No. R3597000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3597000500 500 mL Natural Poly Bottle
R3597005250 250 mL Natural Poly Bottle
3597-32 1 L Natural Poly Bottle
R35970004F 4 L Cubitainer
3597-1 4 L Natural Poly Bottle
3597-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R3597000500 Supplier Ricca Chemical Company Supplier No. 359716
Add to Cart
Add to Cart

Hydrochloric Acid Titrant

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH <2
Color Colorless
Melting Point 0°C
Boiling Point 100°C
CAS Min % 99.82, 0.18
CAS Max % 99.82, 0.18
Density 1.0009
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Solubility Information Miscible in water
Grade Laboratory
Chemical Name or Material Hydrochloric Acid, 0.0500 Normal (N/20)
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.