Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.1% v/v, Reagents

Catalog No. 01335155
Change view
Click to view available options
Quantity:
500 mL
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-155 200 L
01-335-154 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-155 Supplier Reagents Holdings Llc Supplier No. CH001500200E
Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 0.1% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH 3
Color Colorless
Boiling Point 100°C
Packaging Poly Drum
Specific Gravity 1.0002
Quantity 200 L
Physical Form Liquid
Percent Purity 0.1%
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 0.1% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.