Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.1142 N, for Kjeldahl Analysis of Proteins, Ricca Tightrant™
SDP

Catalog No. 36105 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3610-5 20 L Cubitainer
3610-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3610-5 Supplier Ricca Chemical Company Supplier No. 36105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of Proteins (Kjeldahl)

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

Molecular Formula ClH
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 97.0, 1.08
Color Colorless
CAS 7647-01-0, 7732-18-5
Physical Form Liquid
Quantity 20 L
CAS Max % 1.13
Packaging Cubitainer
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1142Normal
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.