Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.12 N in Isopropanol with Bromthymol Blue, Reagents

Catalog No. 01335153
Change view
Click to view available options
:
Quantity: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-153 Quantity: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-153 Supplier Reagents Holdings Llc Supplier No. CH001700100A
Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 0.12 N in Isopropanol with Bromthymol Blue, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hydrochloric Acid, 0.12 N in Isopropanol with Bromthymol Blue
Color Orange
CAS 67-63-0, 7732-18-5
Boiling Point 82°C
Packaging Poly Bottle
Specific Gravity 0.7863
Quantity 100 mL
Molecular Formula HCl/C3H7OH
Physical Form Liquid
Synonym Hydrogen chloride solution
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.