Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hydrochloric Acid Standard Solution, 0.10 N, 1 L

Catalog No. 501995129 Shop All Hach Company Products
Encompass

Multi-purpose hydrochloric acid standard solution, 0.10 N (0.10 M)

Catalog No. 50-199-5129 Supplier Hach Company Supplier No. 1481253
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-purpose hydrochloric acid standard solution, 0.10 N (0.10 M)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.