Learn More
Hydrochloric Acid, 0.250 N (N/4) in Isopropyl Alcohol, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| Concentration | 0.250Normal |
| CAS Min % | 95.0, 2.99 |
| CAS Max % | 99.0, 3.11 |
| Packaging | Cubitainer |
| Solvent or Matrix | Isopropyl Alcohol |
| Quantity | 20 L |
| Physical Form | Liquid |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.