Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1.0334 N, Ricca Chemical
SDP

Catalog No. R37013001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37013001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37013001A Supplier Ricca Chemical Company Supplier No. R37013001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General laboratory reagent for versatile use in analytical and chemical applications.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 36.46 g/mol
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Appearance of Solution Colorless Liquid
Concentration 1.0330 to 1.0338 N
CAS Max % 96.33, 3.67
Packaging Natural Poly Bottle
Quantity 1 L
Grade Not Available
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.