Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Hydrochloric Acid 1 0N 20L UN1789
SDP

Supplier:  Thomas Scientific C748B74

Encompass_Preferred

Hydrochloric Acid 1 0N 20L UN1789

Catalog No. 50-005-50742


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.