Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagents

Catalog No. 01336788
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-788 1 L
01-336-789 10 L
01-336-787 2.5 L
01-336-785 4 L
01-336-784 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-336-788 Supplier Reagents Holdings Llc Supplier No. C55050001A
Add to Cart
Add to Cart

Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH 0
Color Colorless to Slight Yellow
Boiling Point 108°C
Packaging Poly Bottle
Specific Gravity 1.0999
Quantity 1 L
Physical Form Liquid
Percent Purity 50%
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 1:1, 50% v/v Approximately 6N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.