Learn More
StatLab Hydrochloric Acid 10%

Description
- 10% Hydrochloric Acid is used for:
- Iron detection stains
- Tissue decalcification
- Stain differentiation
- StatLab MasterTech special stain kit component
Specifications
Specifications
| Chemical Name or Material | Hydrochloric Acid |
| Concentration | 10% |
| CAS | 7647-01-0 |
| Quantity | 1 L |
| Percent Purity | 10% |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.