Learn More
Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colorless to pale yellow liquid |
Color | Colorless to Yellow |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Quantity | 500 mL |
Physical Form | Liquid |
Solubility Information | Completely miscible with water. |
IUPAC Name | chlorane |
Formula Weight | 36.46 |
Concentration or Composition (by Analyte or Components) | Hydrochloric acid: 10.0%; Water: 90.0% |
Chemical Name or Material | Hydrochloric acid |
Safety and Handling
Hazard Category | H290-H315-H318 |
Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390 |
DOTInformation | Hazard Class: 8, Packing Group: III |
EINECSNumber | 231-595-7 |
RTECSNumber | MW4025000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.