Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrochloric acid, 7647-01-0, MFCD00011324, 100 mL
Catalog No. NC0691401
Encompass
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, 36.5-38.0%, BioReagent, for molecular biology. Hydrochloric acid has been used: for the solubilization of formazan crystals, to adjust pH in various buffers, and for the preparation of giant reed extract.
This item is not returnable. View return policy
Catalog No. NC0691401 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H1758100ML
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, 36.5-38.0%, BioReagent, for molecular biology. Hydrochloric acid has been used: for the solubilization of formazan crystals, to adjust pH in various buffers, and for the preparation of giant reed extract.