Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Hydrochloric Acid
SDP

Catalog No. 21033259
Change view
Click to view available options
Solution Type:
Decalcifier
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solution Type
21-033-259 Decalcifier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-259 Supplier StatLab Supplier No. 3378016
Add to Cart
Add to Cart

Reagent ACS Grade, act as agent in certain staining procedures to remove excess dye and achieve clearer tissue visualization

  • Concentration is 37.4%
  • Used for decalcification of bone tissue by dissolving calcium deposits, allowing for easier sectioning and staining of bone samples
  • Solution is also utilized as a differentiating agent in certain staining procedures to remove excess dye and achieve clearer tissue visualization

Specifications

Product Type Decalcifying Solution
Solution Type Decalcifier
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.