Learn More
Hydrochloric acid, 1N volumetric Standard solution, Macron Fine Chemicals™
Shop All Avantor ProductsSpecifications
Specifications
| Color | Colorless |
| Packaging | Cubitainer |
| Quantity | 4 L |
| Physical Form | Liquid |
| Formula Weight | 36.46 |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric acid, 1N volumetric standard solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.