Learn More
Electron Microscopy Sciences Hydrochloric Acid 2% 500 ML
SDP
Catalog No. 5031866
Encompass_Preferred
Used in Prussian Blue Method for Hemosiderin for the staining of deposits of hemosiderin. For references see Perls, M., Virchow's Arch. F. Path. Anat., 39:42-48, 1867, Highman, B., Arch. Path., 33:937-938, 1942, Lillie, R.D. Histopathologic Technic and Practical Histo-chemistry, 3rd Ed. McGraw-Hill, N.Y., 1965 or Clark, G., (ed), Staining Procedures 3rd Ed., Williams & Wilkins, Baltimore, p. 157, c. 1973.
Catalog No. 50-318-66 Supplier Electron Microscopy Sciences Supplier No. 2661302
Used in Prussian Blue Method for Hemosiderin for the staining of deposits of hemosiderin. For references see Perls, M., Virchow's Arch. F. Path. Anat., 39:42-48, 1867, Highman, B., Arch. Path., 33:937-938, 1942, Lillie, R.D. Histopathologic Technic and Practical Histo-chemistry, 3rd Ed. McGraw-Hill, N.Y., 1965 or Clark, G., (ed), Staining Procedures 3rd Ed., Williams & Wilkins, Baltimore, p. 157, c. 1973.