Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 2 M, Innovating Science™
SCI_ED

Catalog No. S031133 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031133 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031133 Supplier Aldon Corporation Supplier No. IS17019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory-grade hydrochloric acid solution for use in chemical analysis and scientific research.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0

Specifications

Concentration 2 M
Packaging HDPE
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.