Learn More
Aqua Solutions Hydrochloric Acid 2 Normal
SDP
Catalog No. NC1552799
Encompass_Preferred
Composition: 18.517% Hydrochloric Acid (7647-01-0), 0.059% Renex 30 (24938-91-8), 81.425% Water (7732-18-5)
Catalog No. NC1552799 Supplier Aqua Solutions Supplier No. AXI017
Composition: 18.517% Hydrochloric Acid (7647-01-0), 0.059% Renex 30 (24938-91-8), 81.425% Water (7732-18-5)