Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Hydrochloric Acid 20% v/v Solution (4L)
SDP

Catalog No. NC0685834
Encompass

Hydrochloric Acid 20% v/v Solution (4L)

Catalog No. NC0685834 Supplier Aqua Solutions Supplier No. EQS242A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid 20% v/v Solution (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.