Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 30.0%, Suprapur™, MilliporeSigma™

Catalog No. M1003180250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003180250 250 mL
M1003180500 500 mL
M1003181000 1 L
M1003182500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1003180250 Supplier MilliporeSigma Supplier No. 1.00318.0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-01-0 | ClH | 36.46 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH < 1 (300 g/L, H2O, 20°C)
Melting Point −50°C
Boiling Point 85°C (1013 hPa)
Density 1.15 g/cm3 (at 20°C)
Packaging Plastic Bottle
Vapor Pressure 21.8 hPa (20°C)
Assay Percent Range ≥ 30.0%
Quantity 250 mL
Percent Purity 30%
Grade Not Available
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.