Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 33.3% (w/w) with Methyl Orange Indicator, Reagents

Catalog No. 01335037
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-037 4 L
01-335-039 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-037 Supplier Reagents Holdings Llc Supplier No. CH1190004A
Add to Cart
Add to Cart

Hydrochloric Acid, 33.3% (w/w) with Methyl Orange Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Green/Yellow
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.055
Quantity 4 L
Physical Form Liquid
Percent Purity 33.3%
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 33.3% (w/w) with Methyl Orange Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.