Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 35.0 to 39.0%, Solstice

Catalog No. 60047672 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-672 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-672 Supplier Solstice Supplier No. 844361L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tested according to Ph.Eur., fuming 37%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH >0.1 (at 20°C)
Color Colorless
Melting Point ∼-35°C
Boiling Point ∼42°C (1013hPa)
Density 1.2g/mL at 25°C
Odor Stinging
Packaging Glass bottle
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 1 L
Physical Form Liquid
UN Number UN1789
Beilstein 1098214
Solubility Information Completely miscible in water
Formula Weight 36.46g/mol
Percent Purity 35.0 to 39.0%
Grade Reagent
Chemical Name or Material Hydrochloric acid, Tested according to Ph.Eur., fuming 37%
Show More Show Less
Hazard Statement P261-P280-P305 + P351 + P338-P310
DOTInformation 8/PGII
RTECSNumber 231-595-7
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.