Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 36.5-38.0%, BAKER INSTRA-ANALYZED Reagent, For ACS analysis, J.T. Baker™

Catalog No. 14650235 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-650-235 2.5 L
14-650-234 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-650-235 Supplier Avantor Supplier No. 953033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 50°C
Odor Pungent
Residue after Ignition ≤3 ppm
Specific Gravity 1.185 - 1.192 at 60°F
Assay Percent Range 36.5 to 38.0 %
Quantity 2.5 L
Physical Form Liquid
Formula Weight 36.46
Percent Purity 37.7%
Grade Not Available
Chemical Name or Material Hydrochloric Acid, ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.