Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 36%, ACS Grade, Innovating Science™
SCI_ED

Catalog No. S031132 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031132 500 mL
S031134 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S031132 Supplier Aldon Corporation Supplier No. IS17030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Amber Glass
Quantity 500 mL
Molecular Weight (g/mol) 36.46
Percent Purity 36%
Grade ACS
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.