Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 37%, ACROS Organics™

Click to view available options
Quantity:
5L
Packaging:
Plastic Drum
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
chlorane
[H+].[Cl-]
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
chlorane
[H+].[Cl-]

Hydrochloric Acid, 37%

Viscosity 1.9 mPa.s (15°C)
Quantity 5L
Assay Percent Range 37%
Linear Formula HCl
Density 1.1800g/mL
Formula Weight 36.46
Percent Purity 37%
Grade Extra Pure
Packaging Plastic Drum
Solubility Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Specific Gravity 1.18
Melting Point -35.0°C
Boiling Point 57.0°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less