Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid 37% LM Prime, Transene

Catalog No. 502744720
Encompass
Catalog No. 50-274-4720 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T0708312GAL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid, 37 Percent, Electronic/Cleanroom Grade

Hydrochloric acid is a key ingredient to remove surface metallic ions by forming soluble metal chlorides.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Weight (g/mol) 36.5 g/mol

Specifications

Packaging HDPE
Quantity 1 gal.
Percent Purity 37%
Grade Electronic
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.