Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 37%, Trace Metals grade, Ricca Chemical
SDP

Catalog No. RABH002025D Shop All Ricca Chemical Company Products
Change view
Click to view available options
Package: Safety-Coated Glass; Size: 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RABH002025D Package: Safety-Coated Glass; Size: 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RABH002025D Supplier Ricca Chemical Company Supplier No. RABH00202.5D
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

CAS Max % 66, 38
Packaging Safety-coated Glass Bottle
Quantity 2.5 L
Percent Purity 34 to 38%
Grade TraceMetal
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.