Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid 37% w/w, ACS Reagent, Reagents

Catalog No. 01336836
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
Safety Coated Glass
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-836 20 L HDPE Winpak
01-336-840 1 L Glass Bottle
01-336-839 2.5 L Poly Bottle
01-336-838 2.5 L Safety Coated Glass
01-336-837 2.5 L Glass Bottle
01-336-835 4 L Poly Bottle
01-336-834 500 mL Poly Bottle
01-336-833 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-336-836 Supplier Reagents Holdings Llc Supplier No. C510110020E7
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid 37% w/w, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Muriatic Acid, Chlorohydric Acid
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Slightly Greenish-Yellow
Boiling Point 100°C
Packaging HDPE Winpak
Specific Gravity 1.18
Quantity 20 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Hydrochloric Acid 37% w/w, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.