Learn More
HCL Labels Hydrochloric Acid GHS Label - 2" x 3" Adhesive Vinyl (Pack of 25)
SDP
Catalog No. NC0655936
Encompass_Preferred
Hydrochloric Acid 2" x 3" (Pack of 25) GHS Compliant Chemical Safety Label for secondary containers. Made from a dependable 3M adhesive vinyl that is laminated for chemical and solvent resistance. HCL's GHS labels have been tested to withstand acetone, dichloromethane, hydrofluoric acid, and other tough chemicals and solvents. These labels are used for containers that store hazardous substances in order to provide employees with the right-to-know of the immediate chemical hazards. Our GHS labels are compliant with OSHA HazCom standard 1910.1200 and aligned with the Globally Harmonized System for identifying and classifying chemical hazards. HCL's GHS safety labels are made in the United States of America.
Catalog No. NC0655936 Supplier HCL Labels Supplier No. GH105DAVN0023
Add to Cart
Add to Cart
Hydrochloric Acid 2" x 3" (Pack of 25) GHS Compliant Chemical Safety Label for secondary containers. Made from a dependable 3M adhesive vinyl that is laminated for chemical and solvent resistance. HCL's GHS labels have been tested to withstand acetone, dichloromethane, hydrofluoric acid, and other tough chemicals and solvents. These labels are used for containers that store hazardous substances in order to provide employees with the right-to-know of the immediate chemical hazards. Our GHS labels are compliant with OSHA HazCom standard 1910.1200 and aligned with the Globally Harmonized System for identifying and classifying chemical hazards. HCL's GHS safety labels are made in the United States of America.