Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5.0 N Solution, Spectrum™ Chemical
SDP

Catalog No. 18610917 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
55 gal.
Packaging:
Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-917 2.5 L Poly Bottle
18-610-915 1 L Poly Bottle
18-610-916 4 L Poly Bottle
18-610-914 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-917 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1062.5LT
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5

Specifications

Concentration 5.0 N ± 0.05 N
CAS Max % 10 to 20%, 80 to 100%
Packaging Poly Bottle
Quantity 2.5 L
Molecular Weight (g/mol) 36.46
Grade Not Available
Chemical Name or Material Hydrochloric Acid

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.