Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5.00 N, Ricca Chemical
SDP

Catalog No. 374032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3740-32 1 L Natural Poly Bottle
R3740000500 500 mL Natural Poly Bottle
374032G 1 L Amber Glass Bottle
37405PT 2.5 L Safety-coated Glass Bottle
R37400004F 4 L Cubitainer
3740-1 4 L Natural Poly Bottle
R374000010F 10 L Cubitainer
3740-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 3740-32 Supplier Ricca Chemical Company Supplier No. 374032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5 | Formula: ClH | Weight: 36.46 g/mol

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0

Specifications

Packaging Natural Poly Bottle
Quantity 1 L
Molecular Weight (g/mol) 36.46 g/mol
Grade Laboratory
Chemical Name or Material HYDROCHLORIC ACID, 5.00 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.