Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 5% v/v aq. soln.

Catalog No. AA43335K7
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43335K7 4 L
AA43335AP 500 mL
AA43335K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43335K7 Supplier Thermo Scientific Chemicals Supplier No. 043335K7
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7647-01-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula HCl
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym Muriatic acid, Hydrogen chloride, HCl, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 313
ChEBI CHEBI:53434
SMILES Cl

Specifications

Color Colorless to Yellow
Concentration 5% v/v aq. soln.
Quantity 4 L
Physical Form Liquid
UN Number UN1789
IUPAC Name chlorane
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid
Hazard Category H290
Precautionary Statement P234, P390, P406, P501
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber 231-595-7
RTECSNumber MW4025000
TSCA Yes
Recommended Storage Keep containers tightly closed in a dry, cool and well-ventilated place.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.