Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrochloric acid, 7647-01-0, MFCD00011324, 500 mL
Catalog No. NC0695710
Encompass
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, 36.5-38.0%, BioReagent, for molecular biology.
Catalog No. NC0695710 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H1758500ML
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, 36.5-38.0%, BioReagent, for molecular biology.