Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 6.00 Normal with Chloride Testing, Ricca Chemical
SDP

Catalog No. R375200055E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity: 55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R375200055E Quantity: 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R375200055E Supplier Ricca Chemical Company Supplier No. R375200055E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity 6.00 N hydrochloric acid solution formulated for chloride testing applications.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance Colorless to slightly greenish-yellow liquid
pH <1
Packaging 55 Gallon Drum
Specific Gravity 1.1
Normality 6 Normal
Quantity 55 gal.
UN Number UN1789
Grade Not Available
Chemical Name or Material Hydrochloric Acid
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.