Learn More
Hach Company Hydrochloric Acid Digital Titrator Cartridge, 8.00 N
Catalog No. 501995015 Shop All Hach Company Products
Encompass
For base concentration determination by Digital Titrator titration. Digits x 0.01 = meq
Catalog No. 50-199-5015 Supplier Hach Company Supplier No. 1439001
 Restricted Item
Add to Cart
Add to Cart
For base concentration determination by Digital Titrator titration. Digits x 0.01 = meq