Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 99.99% Trace Metals Basis, Solstice

Catalog No. 60046312 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-312 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-312 Supplier Solstice Supplier No. 339253500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

37 wt. % in H2O, 99.999% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH >0.1 (at 20°C)
Color Colorless
Melting Point -35°C
Boiling Point 42°C
Density 1.2g/mL at 25°C
Odor Stinging
Packaging Poly Bottle
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 500 mL
Physical Form Liquid
UN Number UN1789
Beilstein 1098214
Formula Weight 36.46g/mol
Percent Purity 99.999% (trace metals basis)
Grade Not Available
Chemical Name or Material Hydrochloric acid
Show More Show Less
Hazard Statement P261-P280-P305 + P351 + P338-P310
DOTInformation Class 8/ PGII
RTECSNumber 231-595-7
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.