Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hydrochloric Acid, ACS, 12M
SCI_ED

Catalog No. S25838A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
ACS Grade
ACS Grade - SAFE-COTE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25838A 2.5 L ACS Grade - SAFE-COTE
S25358 500 mL ACS Grade - SAFE-COTE
S25838 2.5 L ACS Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25838A
Add to Cart
Edge
Add to Cart

ACS Grade solution designed for educational use. Ideal for laboratory experiments, classroom instruction, and scientific research.

  • ACS Grade Solution
  • Available in 500 mL or 2.5 L size
  • Product is for educational use

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5 , 7647-01-0 , 7732-18-5 , 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Density 1.18
Packaging ACS Grade - SAFE-COTE
Assay Percent Range 36.5% v/v
Quantity 2.5 L
Physical Form Liquid
Linear Formula HCl
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric Acid
ShelfLife 2 years
DOTInformation DOT Class 8, II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.