Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrochloric acid, 7647-01-0, MFCD00011324, 2.5 L
Catalog No. NC0772126
Encompass
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, ACS reagent, 37%. Chlorine source for safer, greener chlorination of aromatic compounds and a,b-unsaturated carbonyl compounds.
Catalog No. NC0772126 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2581482.5L
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46, ACS reagent, 37%. Chlorine source for safer, greener chlorination of aromatic compounds and a,b-unsaturated carbonyl compounds.