Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid ACS AR, Macron Fine Chemicals™

Catalog No. MK5587212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 L
10 lb.
50 lb.
4 x 2.5 L
Packaging:
Glass
Poly
Poly Pail
SAFEMOR
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-5587-212 2.5 L SAFEMOR
MK-H613-10 10 lb. Poly
MK-H613212X4 4 x 2.5 L Glass
MK-H613-50 50 lb. Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MK-5587-212 Supplier Avantor Supplier No. 558746

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging SAFEMOR
Quantity 2.5 L
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.