Learn More
Hydrochloric acid, ACS, HCl 36.5-38.0%
Description
Acidizing petroleum wells, chemical intermediate, ore reduction, food processing, pickling and metal cleaning, general cleaning, and laboratory reagent.Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Appearance | Colorless pungent |
| Color | Colorless |
| Density | 1.19 |
| Odor | Pungent |
| Assay Percent Range | 36.5-38.0% |
| Quantity | 6 lb. |
| Physical Form | Liquid |
| MDL Number | MFCD00011324, MFCD00792839 |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| Formula Weight | 36.46 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.