Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RABH00104C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Amber Glass Bottle
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABH00104C 4 L Amber Glass Bottle
RABH0010500 500 mL Glass Bottle
RABH00101C 1 L Amber Glass Bottle
RABH00102-5 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RABH00104C Supplier Ricca Chemical Company Supplier No. RABH00104C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
CAS Min % 98.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 1 ppm
Packaging Amber Glass Bottle
Residue after Ignition max. 5 ppm
Quantity 4 L
Synonym hydrochloric acid
Percent Purity 36.5 to 38.0%
Grade ACS
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.