Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
4L
Packaging:
Narrow Mouth PTFE Bottle
Poly Bottle
Pressure Bottom Poly Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Quantity 500mL
Formula Weight 36.46
Packaging Poly Bottle
Specific Gravity 1L = 1.18kg
Chemical Name or Material Hydrochloric Acid
Grade BAKER INSTRA-ANALYZED™ Plus