Learn More
Specifications
Specifications
| Color | Colorless |
| Concentration | 0.0500N ±0.0005N (0.05M) |
| CAS Max % | 99.82, 0.18 |
| Density | 1g/mL |
| Identification | Passes Test |
| Packaging | Cubitainer™ |
| Specific Gravity | 1g/mL |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Hydrogen chloride |
| Quantity | 20 L |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.