Learn More
Specifications
Specifications
| Color | Colorless |
| Concentration | 6.25N ±0.05N (6.25M) |
| Density | 1.1g/mL |
| CAS Max % | 79.31, 20.69 |
| Identification | Passes Test |
| Packaging | HDPE Bottle |
| Specific Gravity | 1.1g/mL |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Hydrogen chloride; Thermal decomposition generates: Corrosive vapors |
| Quantity | 2.5 L |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Store in a well-ventilated place. Keep container tightly closed. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.