Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid concentrate, For 1L standard solution, 0.5 M HCl (0.5N), Solstice

Catalog No. 60046716 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-716 6 x 1 Ea.
60-007-70 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-716 Supplier Solstice Supplier No. 382856X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.5 M HCl (0.5N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Boiling Point 100°C (literature)
CAS Min % ≥70.0000%, ≥20.0000%
Concentration 0.5 M HCl (0.5N)
Density 1.012 g/cm3
CAS Max % <80.0000%, <30.0000%
Packaging Ampoule
Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 1 Ea.
Physical Form Concentrate
Linear Formula HCl
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Grade Not Available
Chemical Name or Material Hydrochloric acid concentrate, For 1L standard solution, 0.5 M HCl (0.5N)
Show More Show Less
Hazard Statement P305 + P351 + P338
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Hydrochloric acid
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.